Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122275

Eppendorf® DNA LoBind tubes

capacity 25 mL, PCR clean, screw top cap, withstand 17,000xg, pkg of 200 ea (5 bags x 40 tubes)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
Pricing and availability is not currently available.

material

conical bottom
polypropylene
screw top cap

sterility

non-sterile

feature

DNase free
PCR clean
RNase free
graduations: no
lid

packaging

pkg of 200 ea (5 bags x 40 tubes)

manufacturer/tradename

Eppendorf® 0030122275

parameter

17,000 x g rpm speed

technique(s)

DNA purification: suitable
PCR: suitable

capacity

25 mL

color

colorless

suitability

suitable for PCR
suitable for molecular biology

binding type

DNA low binding

Looking for similar products? Visit Product Comparison Guide

General description

DNA LoBind Tubes, screw cap, DNA LoBind, 25 mL, PCR clean, colorless, 200 tubes (4 bags × 50 tubes)
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 17,000_x_g centrifugation speed for molecular biology applications

Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service