Key Documents
Z691534
MagLab storage system
MagWipe only
Select a Size
1 EA
$50.60
$50.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$50.60
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
$50.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
holds pack of wipers
manufacturer/tradename
Heathrow Scientific HD23502
L × W × H
130 mm × 91 mm × 98 mm
color
green
General description
With lab bench top space at a premium, common lab products such as pipettors, test tubes and miscellanous accessories can be easily organized with Heathrow Scientific′s magnetic lab
storage system, the MagLab. MagLab includes four magnetic pieces: “MagRack” that holds (9) 15ml and (4) 50ml tubes; “Mag Wipe” that holds a standard size (11cm x 21cm) Kimwipes; box; “MagPette” that
holds two pipettors (universal design fits most common brands); and “MagBox” that holds miscellaneous items such as pens/pencils, notes, etc. Each rack is made of robust lightweight ABS plastic in bright and
fun translucent colors and features four powerful magnets for a secure hold onto most metal surfaces or create your own holding point with the metal plate included for each segment.
Legal Information
MagLab is a trademark of Heathrow Scientific
MagWipe is a trademark of Heathrow Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)