Skip to Content
Merck
All Photos(1)

Key Documents

EMU014681

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ezh2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGTTCAGAGGGAGCAAAGCTTGCATTCATTTCATACGCTCTTCTGTCGACGATGTTTTAAGTATGACTGCTTCCTACATCCCTTCCATGCAACACCCAACACATATAAGAGGAAGAACACAGAAACAGCTTTGGACAACAAGCCTTGTGGACCACAGTGTTACCAGCATCTGGAGGGAGCTAAGGAGTTTGCTGCTGCTCTTACTGCTGAGCGTATAAAGACACCACCTAAACGCCCAGGGGGGCGCAGAAGAGGAAGACTTCCGAATAACAGTAGCAGACCCAGCACCCCCACCATCAGTGTGCTGGAGTCAAAGGATACAGACAGTGACAGAGAAGCAGGGACTGAAACTGGGGGAGAGAACAATGATAAAGAAGAAGAAGAGAAAAAAGATGAGACGTCCAGCTCCTCTGAAGCAAATTCTCGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B C Roy et al.
Oncogene, 34(34), 4519-4530 (2014-12-09)
The enhancer of zeste homolog-2 (EZH2) represses gene transcription through histone H3 lysine-27-trimethylation (H3K27me3). Citrobacter rodentium (CR) promotes crypt hyperplasia and tumorigenesis by aberrantly regulating Wnt/β-catenin signaling. We aimed at investigating EZH2's role in epigenetically regulating Wnt/β-catenin signaling following bacterial
Lu Lu et al.
Toxicology and applied pharmacology, 289(2), 276-285 (2015-09-30)
Lung cancer is regarded as the leading cause of cancer-related deaths, and cigarette smoking is one of the strongest risk factors for the development of lung cancer. However, the mechanisms for cigarette smoke-induced lung carcinogenesis remain unclear. The present study
Ying Qi et al.
Molecular bioSystems, 11(7), 1980-1986 (2015-05-08)
A histone methyltransferase enhancer of zeste homologue 2 (EZH2) catalyzes trimethylation at histone H3 lysine27 (H3K27me3) and is frequently dysregulated in a wide range of human cancers. EZH2-mediated gene silencing contributes to carcinogenesis and regulates stem cell maintenance and differentiation;
Jessica Svedlund et al.
Endocrine-related cancer, 21(2), 231-239 (2013-12-03)
Primary hyperparathyroidism (pHPT) resulting from parathyroid tumors is a common endocrine disorder with incompletely understood etiology. In renal failure, secondary hyperparathyroidism (sHPT) occurs with multiple tumor development as a result of calcium and vitamin D regulatory disturbance. The aim of
Gongchao Wang et al.
American journal of cancer research, 5(10), 3162-3173 (2015-12-23)
Lung adenocarcinoma (LAD) and lung squamous cell cancer (LSCC) are two most common histological types of lung cancer, while they differ in many aspects. Recent evidence shows that long non-coding RNAs (lncRNAs) play an important role in the process of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service