Skip to Content
Merck
All Photos(1)

Documents

EHU123831

Sigma-Aldrich

MISSION® esiRNA

targeting human DNMT3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAATGACCTCTCCATCGTCAACCCTGCTCGCAAGGGCCTCTACGAGGGCACTGGCCGGCTCTTCTTTGAGTTCTACCGCCTCCTGCATGATGCGCGGCCCAAGGAGGGAGATGATCGCCCCTTCTTCTGGCTCTTTGAGAATGTGGTGGCCATGGGCGTTAGTGACAAGAGGGACATCTCGCGATTTCTCGAGTCCAACCCTGTGATGATTGATGCCAAAGAAGTGTCAGCTGCACACAGGGCCCGCTACTTCTGGGGTAACCTTCCCGGTATGAACAGGCCGTTGGCATCCACTGTGAATGATAAGCTGGAGCTGCAGGAGTGTCTGGAGCATGGCAGGATAGCCAAGTTCAGCAAAGTGAGGACCATTACTACGAGGTCAAACTCCATAAAGCAGGGCAAAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Nomura et al.
Cells, tissues, organs, 207(3-4), 115-126 (2019-10-02)
Stem cells have essential applications in in vitro tissue engineering or regenerative medicine. However, there is still a need to understand more deeply the mechanisms of stem cell differentiation and to optimize the methods to control stem cell function. In
Xi Han et al.
Cell biology international, 45(1), 227-237 (2020-10-23)
Emerging evidence suggests that miR-143 plays an important role in the regulation of tumor sensitivity to chemotherapeutic agents. The study explores the underlying mechanism of miR-143 in reversing cisplatin resistance in ovarian cancer. The cisplatin-resistant ovarian cancer cell line A2780/CDDP
Phillip L Palmbos et al.
Cancer research, 75(23), 5155-5166 (2015-10-17)
Bladder cancer is a common and deadly malignancy but its treatment has advanced little due to poor understanding of the factors and pathways that promote disease. ATDC/TRIM29 is a highly expressed gene in several lethal tumor types, including bladder tumors
Li Zhang et al.
Cancer cell international, 20, 325-325 (2020-07-24)
Methylation of histone 3 at lysine 9 (H3K9) and DNA methylation are epigenetic marks correlated with genes silencing. The tumor microenvironment significantly influences therapeutic responses and clinical outcomes. The epigenetic-regulation mechanism of the costimulatory factors Tim-3 and galectin-9 in cervical
Anjin Wang et al.
Oncology letters, 21(4), 272-272 (2021-03-16)
Cervical cancer is the second most common gynecological malignancy. Accumulating evidence has suggested that microRNAs (miRNAs) are involved in the occurrence and development of cervical cancer. The present study aimed to investigate the function and underlying molecular mechanism of microRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service