Skip to Content
Merck
All Photos(1)

Key Documents

EHU073481

Sigma-Aldrich

MISSION® esiRNA

targeting human EZH2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCATGCAACACCCAACACTTATAAGCGGAAGAACACAGAAACAGCTCTAGACAACAAACCTTGTGGACCACAGTGTTACCAGCATTTGGAGGGAGCAAAGGAGTTTGCTGCTGCTCTCACCGCTGAGCGGATAAAGACCCCACCAAAACGTCCAGGAGGCCGCAGAAGAGGACGGCTTCCCAATAACAGTAGCAGGCCCAGCACCCCCACCATTAATGTGCTGGAATCAAAGGATACAGACAGTGATAGGGAAGCAGGGACTGAAACGGGGGGAGAGAACAATGATAAAGAAGAAGAAGAGAAGAAAGATGAAACTTCGAGCTCCTCTGAAGCAAATTCTCGGTGTCAAACACCAATAAAGATGAAGCCAAATATTGAACCTCCTGAGAATGTGGAGTGGAGTGGTGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailong Liu et al.
Molecular cancer research : MCR, 15(9), 1275-1286 (2017-05-26)
Medulloblastoma is the most common malignant brain tumor in children. Although accumulated research has suggested that cancer stem-like cells play a key role in medulloblastoma tumorigenesis, the specific molecular mechanism regarding proliferation remains elusive. Here, we reported more abundant expression
Junchao Xue et al.
Biochimica et biophysica acta, 1863(3), 753-763 (2017-01-08)
Circular RNAs (circRNAs), a class of noncoding RNAs generated from pre-mRNAs, participate in regulation of genes. The mechanism for regulation, however, is unknown. Here, to determine if, in human keratinocyte (HaCaT) cells, circular RNAs are involved in arsenite-induced acceleration of
Jiewei Xu et al.
Pathology, research and practice, 215(6), 152374-152374 (2019-04-07)
EZH2 is a core component of the polycomb repressive complex 2 (PRC2), which catalyzes trimethylation of histone H3 lysine 27 (H3K27me3) and promotes carcinogenesis by epigenetically silencing many tumor suppressor genes. Increased EZH2 expression is a marker of advanced and
Wei Dong et al.
American journal of physiology. Cell physiology, 317(2), C253-C261 (2019-01-17)
Myocardial ischemia-reperfusion (I/R) is a common and lethal disease that threatens people's life worldwide. The underlying mechanisms are under intensive study and yet remain unclear. Here, we explored the function of miR-322/503 in myocardial I/R injury. We used isolated rat
Si Sun et al.
Cancer science, 109(6), 1853-1864 (2018-04-10)
Enhancer of zeste homolog 2 (EZH2), which is overexpressed in a wide range of tumors, contributes to ovarian cancer malignancy in several different ways. We aimed to illustrate the role of EZH2 in ovarian cancer cisplatin resistance and to identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service