Skip to Content
Merck
All Photos(1)

Key Documents

EHU054851

Sigma-Aldrich

MISSION® esiRNA

targeting human HDAC6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATGGAAATTCTGGACACCCGCTGGTCCTCAGCTACATCGACCTGTCAGCCTGGTGTTACTACTGTCAGGCCTATGTCCACCACCAGGCTCTCCTAGATGTGAAGAACATCGCCCACCAGAACAAGTTTGGGGAGGATATGCCCCACCCACACTAAGCCCCAGAATACGGTCCCTCTTCACCTTCTGAGGCCCACGATAGACCAGCTGTAGCTCATTCCAGCCTGTACCTTGGATGAGGGGTAGCCTCCCACTGCATCCCATCCTGAATATCCTTTGCAACTCCCCAAGAGTGCTTATTTAAGTGTTAATACTTTTAAGAGAACTGCGACGATTAATTGTGGATCTCCCCCTGCCCATTGCCTGCTTGAGGGGCACCACTACTCCAGCCCAGAAGGAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Yang et al.
Journal of molecular and cellular cardiology, 127, 143-153 (2018-12-26)
Extracellular pH strongly affects cellular metabolism and function. An acidic environment induced under pathological conditions leads to cardiomyocyte injury and dysfunction, but the underlying mechanisms are still poorly understood. Autophagy has been reported as a cytoprotective mechanism that maintains cellular
Mo Cheng et al.
European journal of pharmacology, 840, 1-8 (2018-10-03)
Emerging evidence shows that cytokines such as interleukins (ILs) are involved in the progression and chemoresistance of multiple tumors, including osteosarcoma (OS). Our present study established the doxorubicin (Dox) resistant human OS MG-63 and HOS cells and named them MG-63/Dox
Shigeki Saito et al.
PloS one, 12(10), e0186615-e0186615 (2017-10-19)
Idiopathic pulmonary fibrosis (IPF) is a chronic, progressive and fatal disease. Histone deacetylase 6 (HDAC6) alters function and fate of various proteins via deacetylation of lysine residues, and is implicated in TGF-β1-induced EMT (epithelial-mesenchymal transition). However, the role of HDAC6
Liuqing Xu et al.
Oncotarget, 8(51), 88730-88750 (2017-11-29)
The role of histone deacetylase 6 (HDAC6) in peritoneal fibrosis remains unknown. In this study, we examined the effect of HDAC6 inhibition on the epithelial-mesenchymal transition (EMT) of peritoneal mesothelial cells and development of peritoneal fibrosis. Treatment with tubastatin A
Ophélia Maertens et al.
Cancer discovery, 9(4), 526-545 (2019-02-03)
Although the majority of BRAF-mutant melanomas respond to BRAF/MEK inhibitors, these agents are not typically curative. Moreover, they are largely ineffective in NRAS- and NF1-mutant tumors. Here we report that genetic and chemical suppression of HDAC3 potently cooperates with MAPK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service