Skip to Content
Merck
All Photos(1)

Key Documents

92961

Millipore

Trehalose Disks

suitable for microbiology, Sterile filter paper discs impregnated with trehalose

Synonym(s):

Carbohydrate discs

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 25 DISCS
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 X 25 DISCS
$126.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Quality Level

sterility

sterile

product line

BioChemika

form

disc

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 10 × 25 discs

technique(s)

microbe id | utilization test: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Proteus spp.
Salmonella spp.
Serratia spp.
Shigella spp.
bacteria

General description

The trehalose fermentation test is based on the fermentation ability of trehalose which results in the production of acid as end products, the pH of the medium will drop, and an indicator changes color to red. Some organisms produce additional CO2 gas which can be made visible with a Durham’s tube. The trehalose discs are aseptically added to any sugar-free basal media to test the ability of the organism to hydrolyze trehalose, a non-reducing disaccharide, as the sole source of carbon and energy during fermentation. Diverse organisms like Enterobacteriaceae, Streptococci, and Enterococci can ferment trehalose.

Application

Trehalose Discs are recommended to detect trehalose fermenting bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service