Skip to Content
Merck

Skip To

E5286

Enniatin B1

From Gnomonia errabunda, ≥95% (HPLC), Pdr5p inhibitor

Synonym(s):

2-(N-Methyl-L-isoleucine) Enniatin B, 3-Butan-2-yl-4,10,16-trimethyl-6,9,12,15,18-penta(propan-2-yl)-1,7,13-trioxa-4,10,16-triazacyclooctadecane-2,5,8,11,14,17-hexone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Check Cart for Availability
$286.00

About This Item

Empirical Formula (Hill Notation):
C34H59N3O9
CAS Number:
Molecular Weight:
653.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Quality level:

$286.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Enniatin B1, from Gnomonia errabunda, ≥95% (HPLC)

biological source

Gnomonia errabunda

Quality Level

assay

≥95% (HPLC)

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

storage temp.

−20°C

SMILES string

N1(C(C(=O)OC(C(=O)N(C(C(=O)OC(C(=O)N(C(C(=O)OC(C1=O)C(C)C)C(C)C)C)C(C)C)C(C)C)C)C(C)C)C(CC)C)C

InChI

1S/C34H59N3O9/c1-16-22(12)25-34(43)46-27(20(8)9)30(39)36(14)23(17(2)3)32(41)44-26(19(6)7)29(38)35(13)24(18(4)5)33(42)45-28(21(10)11)31(40)37(25)15/h17-28H,16H2,1-15H3

InChI key

UQCSETXJXJTMKO-UHFFFAOYSA-N

Application

Enniatin B1 has been used as a mycotoxin in the cell viability assay of primary porcine brain capillary endothelial cells (PBCEC) and human brain microvascular capillary cells (HBMEC).[1] It has also been used as an analytical standard in mass spectrometry measurements.

Biochem/physiol Actions

Enniatins are a group of cyclohexadepsipeptide mycotoxins produced by Gnomonia errabuda and several Fusaria species, with phytotoxic, antibiotic, and insecticidal activities. Enniatins function as ionophors by their incorporation into the cellular membrane to form dimeric structures that transport monovalent ions across the membrane (especially the mitochondrial membranes) affecting oxidative phosphorylation uncoupling. It has been demonstrated that enniatins have a cytotoxic effect on human cancer cells. Furthermore, incubation of H4IIE hepatoma cells with enniatins strongly diminished phosphorylation of the ERK (p44/p42). Enniatins B and B1 inhibit the multi-drug resistance transporter Pdr5p from Saccharomyces cerevisiae, indicating their beneficial potential in cases of drug resistant patients.
Enniatins are cyclohexadepsipeptide mycotoxins that have phytotoxic, antibiotic, and insecticidal activities and function as ionophors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E5411E9661E5161
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

solubility

DMSO: 10 mg/mL, ethanol: 10 mg/mL, methanol: 10 mg/mL

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda

biological source

Gnomonia errabunda


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Isabel Krug et al.
PloS one, 13(5), e0197406-e0197406 (2018-05-17)
Enniatins are common contaminants of food and feed and belong to the group of the "emerging" mycotoxins, which are produced by various Fusarium species. Although a wide range of toxic effects, like antibacterial, antifungal, insecticidal and cytotoxic properties, have been
Chemical assessment of mycotoxin contaminants and veterinary residues in Costa Rican animal feed
Leiva A, et al.
International Journal of Food Contamination, 6(1), 5-5 (2019)
Terje Svingen et al.
Environmental toxicology, 32(5), 1658-1664 (2016-09-16)
Mycotoxins are fungi-born metabolites that can contaminate foods through mould-infected crops. They are a significant food/feed-safety issue across the globe and represent a substantial financial burden for the world economy. Moreover, with a changing climate and fungal biota, there is



Global Trade Item Number

SKUGTIN
E5286-1MG04061826690802

Questions

Reviews

No rating value

Active Filters