Skip to Content
Merck
All Photos(1)

Key Documents

A8851

Sigma-Aldrich

Apicidin

≥98% (HPLC), from microbial

Synonym(s):

Cyclo[(2S)-2-amino-8-oxodecanoyl-1-methoxy-L-tryptophyl-L-isoleucyl-(2R)-2-piperidinexcarbonyl]

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$157.00
5 MG
$537.00

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$157.00
5 MG
$537.00

About This Item

Empirical Formula (Hill Notation):
C34H49N5O6
CAS Number:
Molecular Weight:
623.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

microbial

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: ~1 mg/mL

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@]12CCCCN1C(=O)[C@@H](NC(=O)[C@H](Cc3cn(OC)c4ccccc34)NC(=O)[C@H](CCCCCC(=O)CC)NC2=O)C(C)CC

InChI

1S/C34H49N5O6/c1-5-22(3)30-34(44)38-19-13-12-18-29(38)33(43)35-26(16-9-7-8-14-24(40)6-2)31(41)36-27(32(42)37-30)20-23-21-39(45-4)28-17-11-10-15-25(23)28/h10-11,15,17,21-22,26-27,29-30H,5-9,12-14,16,18-20H2,1-4H3,(H,35,43)(H,36,41)(H,37,42)/t22?,26-,27-,29+,30?/m0/s1

InChI key

JWOGUUIOCYMBPV-PYAAAQPJSA-N

General description

Apicidin is a cyclic tetrapeptide fungal metabolite.[1][2]

Application

Apicidin has been used as a histone deacetylase 2 (HDAC2) inhibitor to study its effects on 5-lipoxygenase (5-LO) mRNA expression in cell lines of Mono Mac6.[3] It has also been used to study the effects of toll-like receptor 8 (TLR-8) stimulation and histone modification on the expression of an activator protein 1 [AP-1] family member(Fra-2) and tissue inhibitor of metalloproteinases 1 (TIMP-1) in monocytes.[4]

Biochem/physiol Actions

Apicidin is a potent inhibitor of histone deacetylase (HDAC). It particularlyinhibits histone deacetylase 1 and 3 (HDAC1 and HDAC3).[5] Apicidin exhibits anti-protozoal activity against apicomplexan metabolite produced by parasites.[1] It also possesses anti-proliferative activity against several cancer cell lines. Apicidin shows anti-cancer activity against human acute promyelocytic leukemia cell.[5]
Potent (nM) cell permeable inhibitor of histone deacetylase.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwon, S.H., et al.
The Journal of Biological Chemistry, 18, 2073-2073 (2002)
J S Kim et al.
Biochemical and biophysical research communications, 281(4), 866-871 (2001-03-10)
Apicidin [cyclo(N-O-methyl-L-tryptophanyl-L-isoleucinyl-D-pipecolinyl-l-2-amino-8-oxodecanoyl)], a novel histone deacetylase inhibitor, has been identified as an antiprotozoal and antiproliferative agent. In this study, we show apicidin induces transcriptional activation of p21(WAF1/CIP1) (p21) in human prostate carcinoma cells. Apicidin induces expression of p21 protein and
Beatrix Meltzer et al.
Virology, 518, 241-252 (2018-03-20)
In HIV infected macrophages, a large population of viral genomes persists as the unintegrated form (uDNA) that is transcriptionally active. However, how this transcriptional activity is controlled remains unclear. In this report, we investigated whether Tat, the viral transactivator of
J W Han et al.
Cancer research, 60(21), 6068-6074 (2000-11-21)
Apicidin [cyclo(N-O-methyl-L-tryptophanyl-L-isoleucinyl-D-pipecolinyl -L-2-amino-8-oxodecanoyl)] is a fungal metabolite shown to exhibit antiparasitic activity by the inhibition of histone deacetylase (HDAC). In this study, we evaluated apicidin as a potential antiproliferative agent. Apicidin showed a broad spectrum of antiproliferative activity against various
Jeong Gu Kang et al.
Scientific reports, 9(1), 11960-11960 (2019-08-21)
Despite the increased interest in epigenetic research, its progress has been hampered by a lack of satisfactory tools to control epigenetic factors in specific genomic regions. Until now, many attempts to manipulate DNA methylation have been made using drugs but

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service