Skip to Content
Merck
All Photos(2)

Key Documents

Z2125

Sigma-Aldrich

Zearalenone

fungal mycotoxin

Synonym(s):

F-2 toxin

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$231.00
25 MG
$431.00

$231.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$231.00
25 MG
$431.00

About This Item

Empirical Formula (Hill Notation):
C18H22O5
CAS Number:
Molecular Weight:
318.36
Beilstein:
1350216
MDL number:
UNSPSC Code:
12191503
PubChem Substance ID:
NACRES:
NA.77

$231.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

storage temp.

−20°C

SMILES string

C[C@H]1CCCC(=O)CCC\C=C\c2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H22O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,20-21H,2,4-6,8-9H2,1H3/b7-3+/t12-/m0/s1

InChI key

MBMQEIFVQACCCH-QBODLPLBSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Zearalenone, a fungal mycotoxin produced by Fusarium, binds the estrogen receptor (ER) and is uterotropic in the newborn rat. It is a common contaminant in cereal grain used for animal and human food, and exerts an estrogenic activity that modulates/disrupts endocrine function in animals and possibly humans.[1]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Leffers et al.
Human reproduction (Oxford, England), 16(5), 1037-1045 (2001-05-02)
We have compared the oestrogenic potency of the synthetic oestrogen Zeranol, used as a growth promoter in meat production, and five related compounds, with the potency of 17beta-oestradiol, diethylstilboestrol (DES), genistein, and Bisphenol-A. The potency was assayed by analysing differences
Katarzyna Cieplińska et al.
Toxins, 11(5) (2019-05-30)
Zearalenone is a frequent contaminant of cereals and their by-products in regions with a temperate climate. This toxic molecule is produced naturally by Fusarium fungi in crops. The aim of this study was to determine the influence of low zearalenone
D M Sheehan et al.
Teratology, 29(3), 383-392 (1984-06-01)
Fusarium sp. contaminated feedstuffs elicit adverse estrogenic effects in several commercially important animal species via the mycotoxin zearalenone. An estrogenically active synthetic derivative, zearalanol, is used as an anabolic agent in cattle. Since estrogens can irreversibly alter target tissue development
Kazimierz Obremski et al.
Toxins, 12(6) (2020-05-31)
The aim of the present study was to determine the effect of zearalenone (ZEN), administered per os to gilts at doses equivalent to 50%, 100%, and 150% of no-observed-adverse-effect level (NOAEL) values for 14, 28, and 42 days during weaning
S De Baere et al.
Analytica chimica acta, 756, 37-48 (2012-11-28)
A sensitive and specific method for the quantitative determination of zearalenone (ZEN) and its major metabolites (α-zearalenol (α-ZEL), β-zearalenol (β-ZEL), α-zearalanol (α-ZAL), β-zearalanol (β-ZAL) and zearalanone (ZAN)) in animal plasma using liquid chromatography combined with heated electrospray ionization (h-ESI) tandem

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service