Skip to Content
Merck
All Photos(2)

Documents

E4765

Sigma-Aldrich

trans-9,10-Epoxystearic acid

~99% (capillary GC)

Synonym(s):

trans-9,10-Epoxyoctadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O3
CAS Number:
Molecular Weight:
298.46
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99% (capillary GC)

form

solid

functional group

carboxylic acid
epoxy

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCC1OC1CCCCCCCC(O)=O

InChI

1S/C18H34O3/c1-2-3-4-5-7-10-13-16-17(21-16)14-11-8-6-9-12-15-18(19)20/h16-17H,2-15H2,1H3,(H,19,20)

InChI key

IMYZYCNQZDBZBQ-UHFFFAOYSA-N

Packaging

Sealed ampule

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 9,10-epoxystearate ester in intravenous solutions stored in poly(vinyl chloride) bags, using gas chromatography-single-ion monitoring mass spectrometry.
G A Ulsaker et al.
The Analyst, 109(7), 967-968 (1984-07-01)
P Fahlstadius
Lipids, 23(11), 1015-1018 (1988-11-01)
Racemic cis-9,10-epoxystearic acid was isolated from total lipids of human leukocytes. Identification of the epoxy acid was based mainly on gas liquid chromatographic-mass spectrometric analysis and on its chemical conversion into threo-9,10-dihydroxystearic acid. A mass spectrometric method for quantitative determination
F Müller et al.
European journal of biochemistry, 245(2), 490-496 (1997-04-15)
Mammalian soluble and microsomal epoxide hydrolases have been proposed to belong to the family of alpha/beta-hydrolase-fold enzymes. These enzymes hydrolyse their substrates by a catalytic triad, with the first step of the enzymatic reaction being the formation of a covalent
F Pinot et al.
Biochemical and biophysical research communications, 184(1), 183-193 (1992-04-15)
A microsomal fraction from etiolated Vicia sativa seedlings incubated aerobically with [1-14C]oleic acid (Z9-octadecenoic acid) or [1-14C]9,10-epoxystearic acid or [1-14C]9,10-dihydroxystearic acid catalyzed the NADPH-dependent formation of hydroxylated metabolites. The chemical structure of compounds formed from oleic, 9,10-epoxystearic or 9,10-dihydroxystearic acids
Alejandro González-Benjumea et al.
Frontiers in bioengineering and biotechnology, 8, 605854-605854 (2021-01-21)
Epoxides of vegetable oils and free and methylated fatty acids are of interest for several industrial applications. In the present work, refined rapeseed, sunflower, soybean, and linseed oils, with very different profiles of mono- and poly-unsaturated fatty acids, were saponified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service