Skip to Content
Merck
All Photos(1)

Key Documents

E5641

Sigma-Aldrich

11,12-Epoxy-(5Z,8Z,14Z)-eicosatrienoic acid

~100 μg/mL in ethanol, ≥95%

Synonym(s):

(±)11,12-EpETrE, 11,12-EET

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O3
CAS Number:
Molecular Weight:
320.47
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

concentration

~100 μg/mL in ethanol

lipid type

omega FAs

storage temp.

−20°C

SMILES string

CCCCC\C=C\CC1OC1C\C=C\C\C=C\CCCC(O)=O

InChI

1S/C20H32O3/c1-2-3-4-5-9-12-15-18-19(23-18)16-13-10-7-6-8-11-14-17-20(21)22/h6,8-10,12-13,18-19H,2-5,7,11,14-17H2,1H3,(H,21,22)/b8-6+,12-9+,13-10+

InChI key

DXOYQVHGIODESM-ATELOPIESA-N

Packaging

Packaged under argon

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yindi Ding et al.
The Journal of pharmacology and experimental therapeutics, 350(1), 14-21 (2014-04-26)
Cytochrome P450-derived epoxides of arachidonic acid [i.e., the epoxyeicosatrienoic acids (EETs)] are important lipid signaling molecules involved in the regulation of vascular tone and angiogenesis. Because many actions of 11,12-cis-epoxyeicosatrienoic acid (EET) are dependent on the activation of protein kinase
Yong Zhou et al.
Shock (Augusta, Ga.), 47(5), 638-645 (2016-10-19)
Acute lung injury (ALI) is characterized by rapid alveolar injury, vascular leakage, lung inflammation, neutrophil accumulation, and induced cytokines production leading to lung edema. The mortality rate of patients suffering from ALI remains high. Epoxyeicosatrienoic acids (EETs) are cytochrome P450-dependent
L Yang et al.
British journal of clinical pharmacology, 80(1), 28-44 (2015-02-07)
There is increasing evidence suggesting that epoxyeicosatrienoic acids (EETs) play an important role in cardioprotective mechanisms. These include regulating vascular tone, modulating inflammatory responses, improving cardiomyocyte function and reducing ischaemic damage, resulting in attenuation of animal models of cardiovascular risk
Xiaojing Wang et al.
Scandinavian journal of immunology, 90(3), e12791-e12791 (2019-05-28)
The epoxyeicosatrienoic acids (EETs) are products of cytochrome P450 epoxygenases and have recently been found to have an anti-inflammatory activity. However, the role of EETs in non-alcoholic steatohepatitis has not been fully understood. In this study, we investigated the protective
Andrea E Vasconez et al.
Acta physiologica (Oxford, England), 225(1), e13168-e13168 (2018-08-05)
The histone demethylase Jarid1b limits gene expression by removing the active methyl mark from histone3 lysine4 at gene promoter regions. A vascular function of Jarid1b is unknown, but a vasoprotective function to inflammatory and hypertrophic stimuli, like angiotensin II (AngII)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service