Skip to Content
Merck
All Photos(1)

Key Documents

D2321

Sigma-Aldrich

DAF-FM DA solution

5 mM in DMSO, ≥97% (HPLC)

Synonym(s):

4-Amino-5-methylamino- 2′,7′-difluorofluorescein diacetate, Diaminofluorescein-FM diacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$925.00

$925.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$925.00

About This Item

Empirical Formula (Hill Notation):
C25H18F2N2O7
CAS Number:
Molecular Weight:
496.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$925.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

DMSO solution

storage condition

desiccated
protect from light

color

clear colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

Fc1cc2c(cc1OC(=O)C)Oc3c(cc(c(c3)OC(=O)C)F)C42OC(=O)c5c4ccc(c5N)NC

InChI

1S/C25H18F2N2O7/c1-10(30)33-20-8-18-13(6-15(20)26)25(12-4-5-17(29-3)23(28)22(12)24(32)36-25)14-7-16(27)21(34-11(2)31)9-19(14)35-18/h4-9,29H,28H2,1-3H3

InChI key

BEVHTVRRVVEMEF-UHFFFAOYSA-N

Biochem/physiol Actions

DAF-FM DA is a cell permeable fluorescent probe for the detection of nitric oxide (NO).
DAF-FM DA is a cell-permeable fluorescent probe for the detection of nitric oxide (NO). DAF-2 is the original compound and is widely used, but DAF-FM and DAF-FM DA are more sensitive to nitric oxide (NO) compared with DAF-2 and more photo-stable and less pH sensitive. DAF-FM DA permeates well into living cells and is rapidly transformed into water-soluble DAF-FM by cytosolic esterases.

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Light sensitive

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurora Parlati et al.
Frontiers in plant science, 8, 1299-1299 (2017-08-10)
The PII protein in plants has been associated to many different tissue specialized roles concerning the Nitrogen assimilation pathways. We report here the further characterization of
Gen Li et al.
Frontiers in plant science, 11, 1215-1215 (2020-09-10)
Anthocyanin-derived fleshy fruit pigmentation has become an excellent system for studying the regulatory network underlying fruit ripening and quality. The transcriptional control of anthocyanin biosynthesis by MYB-bHLH-WDR complexes has been well established, but the intermediate signals through which the environmental
Diana Passaro et al.
Cancer cell, 32(3), 324-341 (2017-09-06)
The biological and clinical behaviors of hematological malignancies can be influenced by the active crosstalk with an altered bone marrow (BM) microenvironment. In the present study, we provide a detailed picture of the BM vasculature in acute myeloid leukemia using
Caihua Wang et al.
Stem cell research & therapy, 10(1), 175-175 (2019-06-15)
Cancer stem cells (CSCs) play a critical role in tumor development and progression and are involved in cancer metastasis. The role of reactive oxygen species (ROS) in CSCs and cancer metastasis remains controversial. The aim of the present study was

Articles

We offer many products related to nitric oxide synthases for your research needs.

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service