Skip to Content
Merck

Skip To

D2196

DAF-FM solution

5 mM in DMSO, ≥97% (HPLC)

Synonym(s):

4-Amino-5-methylamino- 2′,7′-difluorofluorescein, Diaminofluorescein-FM

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
$960.00

About This Item

Empirical Formula (Hill Notation):
C21H14F2N2O5
CAS Number:
Molecular Weight:
412.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
DMSO solution
Storage condition:
desiccated

$960.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

DMSO solution

storage condition

desiccated

color

colorless to yellow, including pink

shipped in

wet ice

storage temp.

−20°C

SMILES string

Fc1cc2c(cc1O)Oc3c(cc(c(c3)O)F)C42OC(=O)c5c4ccc(c5N)NC

InChI

1S/C21H14F2N2O5/c1-25-13-3-2-8-18(19(13)24)20(28)30-21(8)9-4-11(22)14(26)6-16(9)29-17-7-15(27)12(23)5-10(17)21/h2-7,25-27H,24H2,1H3

InChI key

DIJCILWNOLHJCG-UHFFFAOYSA-N

Application

DAF-FM solution has been used as a fluorescent probe for detecting nitric oxide (NO) from plant samples.

Biochem/physiol Actions

DAF-FM is a highly sensitive, photo-stable fluorescent probe for the detection of nitric oxide (NO).
DAF-FM is a highly sensitive, photo-stable fluorescent probe for the detection of nitric oxide (NO). DAF-2 is the original compound and is widely used, but DAF-FM is more sensitive to nitric oxide (NO) compared with DAF-2. DAF-FM (Ex: 500 nm; Em: 515 nm; dectection limit: 3 nM) is more photo-stable and less pH sensitive than DAF-2. DAF-FM is not quite as cell-permeable as the diacetate derivative, but it does not need to be activated by cytosolic enzymes.

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D2321D1821D1946
form

DMSO solution

form

DMSO solution

form

powder

form

powder

assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

desiccated

storage condition

desiccated, protect from light

storage condition

desiccated

storage condition

desiccated

color

colorless to yellow, including pink

color

clear colorless

color

white to brown

color

colorless to yellow


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters