Skip to Content
Merck

Skip To

D225

4,5-Diaminofluorescein diacetate solution

5 mM in DMSO, ≥97% (HPLC)

Synonym(s):

DAF-2 DA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Check Cart for Availability
$1,040.00

About This Item

UNSPSC Code:
41116100
NACRES:
NA.28
MDL number:

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solution

availability

not available in Japan

concentration

5 mM in DMSO

shipped in

wet ice

storage temp.

−20°C

General description

4,5-Diaminofluorescein diacetate (DAF-2 DA) solution is a membrane-permeable fluorescent probe, which measures nitric oxide (NO) in living cells. It is used to study the synthesis of NO in bovine aortic endothelial cells (BAECs). DAF-2 DA imaging shows that ciliated epithelial cells produces high amount of NO than non-epithelial inflammatory cells.

Application

Cell-permeable fluorescent detector of nitric oxide in living cells.

Legal Information

DAF-2 is manufactured by Daiichi Pure Chemicals Co., LTD.
US Patent No. 5,874,590

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D2813D224251505-M
concentration

5 mM in DMSO

concentration

-

concentration

5 mM in DMSO

concentration

-

form

solution

form

lyophilized powder

form

solution

form

solution

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.


Determinants of shear stress-stimulated endothelial nitric oxide production assessed in real-time by 4, 5-diaminofluorescein fluorescence
Qiu W, et al.
Biochemical and Biophysical Research Communications, 286(2), 328-335 (2001)
Increased nitric oxide production in nasal epithelial cells from allergic patients-RT-PCR analysis and direct imaging by a fluorescence indicator: DAF-2 DA
Takeno S, et al.
Clinical and Experimental Allergy, 31(6), 881-888 (2001)
Rachel A Battaglia et al.
eLife, 8 (2019-11-05)
Alexander disease (AxD) is a fatal neurodegenerative disorder caused by mutations in glial fibrillary acidic protein (GFAP), which supports the structural integrity of astrocytes. Over 70 GFAP missense mutations cause AxD, but the mechanism linking different mutations to disease-relevant phenotypes



Global Trade Item Number

SKUGTIN
D225-1MG04061833562017

Questions

Reviews

No rating value

Active Filters