Skip to Content
Merck
All Photos(1)

Key Documents

D157406

Sigma-Aldrich

2-Dimethylaminoethanol

Synonym(s):

N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1209235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

100 mmHg ( 55 °C)
6.12 mmHg ( 20 °C)

Quality Level

refractive index

n20/D 1.4294 (lit.)

bp

134-136 °C (lit.)

mp

−70 °C (lit.)

density

0.886 g/mL at 20 °C (lit.)

SMILES string

CN(C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Sommerfeld
Phytomedicine : international journal of phytotherapy and phytopharmacology, 14(11), 711-715 (2007-10-10)
Tricutan is a combination product of herbal extracts traditionally used for treatment of skin conditions, together with dimethylaminoethanol. The effectiveness of Tricutan in improving skin firmness and elasticity in photoaged facial skin was examined in a randomised, placebo-controlled, double-blind, split-face
Olivier Blin et al.
Psychopharmacology, 207(2), 201-212 (2009-09-17)
Dimethylaminoethanol pyroglutamate (DMAE p-Glu) is a compound resulting from the reaction between dimethylaminoethanol (an indirect precursor of acetylcholine) and pyroglutamic acid (a cyclic derivative of glutamic acid having procholinergic properties and promnesic effects in both animals and man). The present
Tadashi Tatsumi et al.
Bioorganic & medicinal chemistry, 18(7), 2720-2727 (2010-03-17)
Effects of retro-inverso (RI) modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster backbone were clarified. Construction of the isoster backbone was achieved by a stereoselective aldol reaction. Four diastereomers with different configurations at the isoster hydroxyl site and the
K M Houston et al.
Parasitology, 135(Pt 1), 55-61 (2007-09-26)
Juvenile female Litomosoides sigmodontis secrete a protein (Juv-p120) highly modified with dimethylethanolamine (DMAE). In an attempt to establish the source of this decoration worms were pulsed with [3H]-choline and [3H]-ethanolamine and the radio-isotope labelled products analysed. Both isotope labels were
Bruno Dubois et al.
Journal of Alzheimer's disease : JAD, 29(3), 527-535 (2012-02-15)
New criteria related to prodromal Alzheimer's disease (AD) have been proposed to overcome the issue of heterogeneity of patients with mild cognitive impairment (MCI) and to better define patients in early stage AD. Only few therapeutic trials, if any, have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service