Merck
  • Evaluation of retro-inverso modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster.
Evaluation of retro-inverso modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster.
Bioorganic & medicinal chemistry (2010-03-17)
Tadashi Tatsumi, Chiyuki Awahara, Hiromi Naka, Saburo Aimoto, Hiroyuki Konno, Kazuto Nosaka, Kenichi Akaji
ABSTRACT
Effects of retro-inverso (RI) modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster backbone were clarified. Construction of the isoster backbone was achieved by a stereoselective aldol reaction. Four diastereomers with different configurations at the isoster hydroxyl site and the scissile site substituent were synthesized. Inhibitory activities of the new inhibitors suggest that partially modified RI inhibitors would interact with HTLV-1 protease in the same manner as the parent hydroxyethylamine inhibitor.
Sigma-Aldrich
2-Dimethylaminoethanol, purified by redistillation, ≥99.5%
Sigma-Aldrich
2-Dimethylaminoethanol, ≥99.5%