Skip to Content
Merck
All Photos(2)

Key Documents

423041

Sigma-Aldrich

2-Dimethylamino-2-methylpropanol solution

80% in H2O, technical grade

Synonym(s):

DMAMP-80

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$64.90

$64.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$64.90

About This Item

Linear Formula:
(CH3)2C(CH2OH)N(CH3)2
CAS Number:
Molecular Weight:
117.19
Beilstein:
1732837
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$64.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

form

liquid

concentration

80% in H2O

refractive index

n20/D 1.44

bp

61 °C/20 mmHg

density

0.91 g/mL at 25 °C

functional group

amine
hydroxyl

SMILES string

CN(C)C(C)(C)CO

InChI

1S/C6H15NO/c1-6(2,5-8)7(3)4/h8H,5H2,1-4H3

InChI key

XRIBIDPMFSLGFS-UHFFFAOYSA-N

General description

2-Dimethylamino-2-methylpropanol solution (DMAMP-80) is a propanol amine derivative. It is applicable as synthetic intermediate, emulsifying amine and vapor phase corrosion inhibitor. It shows low volatility and higher catalytic activity which makes it a viable alternative to DMEA (dimethylethanolamine) owing in the foam formulation process. It is a non-lachrymator, shows low toxicity profile and has been approved by FDA for indirect food contact.[1] 2-Dimethylamino-2-methyl-1-propanol (DMAMP) has been reported to show good antimicrobial properties against a mixed flora of fungi and bacteria in cutting fluids.[2] The photocatalytic degradation of DMAMP in the presence of TiO2 particles and UV-A (λ = 365nm) radiation has been investigated.[3]

Legal Information

DMAMP-80 is a trademark of Angus Chemie GmbH

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

152.6 °F - Tag closed cup

Flash Point(C)

67 °C - Tag closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chung-Shin Lu et al.
Journal of hazardous materials, 165(1-3), 306-316 (2008-11-18)
The present study deals with the photocatalytic degradation of the alkanolamine, 2-dimethylamino-2-methyl-1-propanol (DMAMP), in the presence of TiO(2) particles and UV-A (lambda=365 nm) radiation. The obtained results show complete oxidation of DMAMP after 20h, and a little over 90% of
Api Api
API Polyurethanes Expo 2001, 552-554 (2001)
Antimicrobial properties of butanolamines and propanolamines in metal working fluids.
Bennett EO, et al.
The Journal of General and Applied Microbiology, 25(2), 63-69 (1979)
M Sandin et al.
Antimicrobial agents and chemotherapy, 34(3), 491-493 (1990-03-01)
The antimicrobial effects of diethanolamine, dimethylamino-methyl-propanol, and butylethanolamine are greatly enhanced at high pH. Their antimicrobial activities are closely correlated with their uncharged forms, indicating that diffusion through cell membrane(s) is rate limiting for the antimicrobial action. Since these compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service