CLS7605
Corning® Transtar® liquid handling system
adjustable volume pipettor
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
sterility
non-sterile
manufacturer/tradename
Corning 7605
volume
200 μL
General description
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Legal Information
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)