Documents
Z370797
Sigma-Aldrich
Vaccu-Pette/96 multiwell pipetter
polystyrene
Synonym(s):
96 well plate pipetting device, caviar dropper, multiwell pipettor
About This Item
UNSPSC Code:
41121500
NACRES:
NB.22
material
polystyrene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37876-0000
L × W × diam.
127 mm × 86 × 37 mm
General description
A simple idea that saves a lot of time in a variety of multiwell protocols. For transfers to, from, and between 96 well plates, just connect the Vaccu-Pette/96 to a large syringe and aspirate about 100 times the desired per-well volume. For example, to transfer 50 μL/well, aspirate and deliver a total of 5 mL. Cells growing in the wells are protected by ridges that prevent access to the bottom 50 μl of each well. Ideal for ELISA, cytotoxicity, and cloning procedures. Without a syringe, the device will pick up about 15 μL per well, making it an effective 96 well blotting tool. 35 mL syringe not included.
Legal Information
Vaccu-Pette/96 is a trademark of SP Scienceware, a division of SP