Skip to Content
Merck
All Photos(2)

Key Documents

M2531

Multiwell plate washer/dispenser manifolds

8 position, bent

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$678.00

$678.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
1 EA
$678.00

About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

$678.00


Estimated to ship onMay 26, 2025


sterility

non-sterile

color

none

Looking for similar products? Visit Product Comparison Guide

General description

For use with 96 well multiwell plates
  • Dispense or aspirate media, or wash, a full row or column at a time
  • Luer fitting
  • Stainless steel and PTFE construction for easy cleaning and autoclavability
  • Optional repeat kit provides continuous operation capability

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service