CLS431685
Corning® Erlenmeyer flask, 5 L
plain bottom, polycarbonate, sterile
Synonym(s):
Corning flask
About This Item
Recommended Products
material
flat bottom flask
polycarbonate
polypropylene cap
vent cap
wide-mouth
wide-mouth neck
sterility
sterile
feature
cap
closure type Vented cap
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Corning 431685
capacity
5 L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Graduated, with a plain bottom
- Sterilized, with sterility assurance level (SAL) of 10⁻⁶
- Nonpyrogenic
- RNase-/DNase-free
- Wide mouth with vented orange polypropylene cap
- Disposable
Other Notes
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service