Skip to Content
Merck
All Photos(1)

Documents

CLS11501

Corning® Erlenmeyer flask, 5 L

plain bottom, with aseptic connections, polycarbonate, sterile, case of 2 ea

Synonym(s):

5-liter Erlenmeyer flask, Corning flask, Laboratory Erlenmeyer flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.43

material

plain bottom
polycarbonate
polypropylene cap
vent cap
wide-mouth neck

sterility

sterile

feature

cap

packaging

case of 2 ea

manufacturer/tradename

Corning 11501

flask capacity

(5,000 mL)

Looking for similar products? Visit Product Comparison Guide

General description

Flask contains a preassembled aseptic transfer cap having two ports: one with 0.2 m polyvinylidene fluoride filter at the end, and the other port with flexible tubing that can be heat-sealed and is chemical resistant, that terminates in a male Luer lock, or a male polycarbonate fast connect.

Application

Corning® Erlenmeyer flask has been used as:
  • transfer medium in liquid handling processes
  • storing and shaking culture application
  • scaling up suspended culture

Features and Benefits

  • Graduated polycarbonate flask with plain bottom, provides a full aseptic solution
  • For efficient transfer of sterile liquid, the dip tube with male plastic coupler (MPC) tubing connector, extends to the bottom
  • Double-bagged with sterility assurance level (SAL) of 10⁻⁶

Other Notes

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service