Documents
CLS431328
Corning® CellBIND® Surface cell culture flasks
CellBIND 175cm2 angled neck cell culture flask w/ Vent Cap and Bar Code, PS, sterile, 84/cs
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
material
angled neck
cap (vented)
high-density polyethylene cap
rectangular flask
rectangular polystyrene
sterility
sterile; γ-irradiated
feature
graduations
packaging
pack of 7
case of 84
manufacturer/tradename
Corning 431328
maximum volume
250 mL
surface area
175 cm2
working volume
35-52.5 mL
suitability
suitable for (cell culture applications; optimal for adherent cell culturing)
General description
The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.
CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
  • Manufactured from optically clear virgin polystyrene.
  • Treated CellBIND surface treatment for enhanced cell attachment
  • 100% integrity tested.
  • Sterilized by gamma irradiation and certified nonpyrogenic
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.