Skip to Content
Merck
All Photos(1)

Key Documents

122-20

Sigma-Aldrich

Rat Collagen Type I

from rat tail, liquid, 0.05 mg/mL, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

20 ML
$59.00

$59.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 ML
$59.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

$59.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Rat Tail Collagen Coating Solution (20 ML), Type I, 50 μg/mL, solution, BioReagent, suitable for cell culture

Quality Level

sterility

sterile; sterile-filtered

product line

BioReagent

form

solution

shelf life

6 mo.

packaging

pkg of 20 mL

concentration

50 μg/mL

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

Collagen type I is a fibrillar collagen that consists of two α1 chains and one α2 chain twisted together to form a triple helix structure. It is the predominant protein in vertebrates and plays a significant role in the extracellular matrices present in various tissues such as bones, tendons, skin, ligaments, cornea, and other types of connective tissue between cells. Rat Tail Collagen Coating Solution (20 ML) enhances cell attachment and proliferation, or promotion of cell-specific morphology and function. It is commonly used for endothelial cells, hepatocytes, muscle cells, and a variety of other cell types.
This collagen solution is prepared in 0.02 M Acetic Acid at 50 ug/ml and sterile filtered.

Application

Rat Tail Collagen Coating Solution (20 ML) is suitable for use in cell culture to study the growth, differentiation, and migration of cells. The rat-tail collagen I coating solution has been used to coat 96-well plates, providing a suitable surface for the adhesion and growth of primary keratinocytes in cell culture experiments.[1]

Preparation Note

To prepare the tissue culture ware, apply Rat Tail Collagen Solution at a culture surface ratio of 5 ug/cm2. Here are some examples:
  • For a T-25 flask, use 125 ug (2.5 ml) of the solution.
  • For each well of a 24-well plate, use 10.5 ug (210 ul) of the solution.
After applying the solution, incubate the coated tissue culture ware at 37oC for 2 hours. Carefully remove the Collagen Coating Solution by aspiration. Rinse the coated surface twice with PBS. The coated tissue culture ware can be used immediately or air-dried and stored at 4oC for up to one week.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariano Maffei et al.
Scientific reports, 9(1), 19214-19214 (2019-12-18)
Gene delivery using vector or viral-based methods is often limited by technical and safety barriers. A promising alternative that circumvents these shortcomings is the direct delivery of proteins into cells. Here we introduce a non-viral, ligand-mediated protein delivery system capable

Questions

  1. Need to dilute with acetic acid or we can use directly

    1 answer
    1. 122-20 is provided ready to use at a concentration of 50ug/mL. No additional dilution is recommended unless required by your application.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service