Skip to Content
Merck
All Photos(1)

Key Documents

CLS3290

Corning® CellBIND® Surface cell culture flasks

CellBIND 75cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 100/cs

Synonym(s):

cell culture flasks, tissue culture flasks

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$59.20
100 EA
$746.00

$59.20

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025



Select a Size

Change View
5 EA
$59.20
100 EA
$746.00

About This Item

UNSPSC Code:
41122104
eCl@ss:
32040501
NACRES:
NB.13

$59.20

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


material

U-bottom flask
canted neck
cap (vented)
high-density polyethylene cap
rectangular flask
rectangular polystyrene
vent cap

sterility

sterile; γ-irradiated

feature

cap
graduations

packaging

case of 100
pack of 5

manufacturer/tradename

Corning 3290

maximum volume

60 mL

surface area

75 cm2

working volume

15-22.5 mL

suitability

suitable for (cell culture applications; optimal for adherent cell culturing)

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.

CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.

Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.

  • Manufactured from optically clear virgin polystyrene.
  • Treated CellBIND surface treatment for enhanced cell attachment
  • 100% integrity tested.
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Caroline Hochheuser et al.
Cancers, 12(11) (2020-11-06)
Background: The bone marrow (BM) is the main site of metastases and relapse in patients with neuroblastoma (NB). BM-residing mesenchymal stromal cells (MSCs) were shown to promote tumor cell survival and chemoresistance. Here we characterize the MSC compartment of the
Karen Peynshaert et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 150, 87-95 (2020-03-17)
Neuroprotection is a mutation-independent therapeutic strategy that seeks to enhance the survival of neuronal cell types through delivery of neuroprotective factors. The Müller cell, a retinal glial cell type appreciated for its unique morphology and neuroprotective functions, could be regarded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service