Skip to Content
Merck
All Photos(1)

Documents

CLS431121

Corning® cryogenic boxes, racks & trays

Box for 100 cryogenic vials, 1-2 mL

Synonym(s):

cryovials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.13

sterility

non-sterile

feature

Holds 100 x 1-2 mL cryovials

packaging

pack of 10 ea

manufacturer/tradename

Corning 431121

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Reusable orange polycarbonate cryogenic vial storage box with lid
  • Box can withstand temperature from - 196°C to +121°C
  • Recommended for use with internally threaded cryogenic vials only
  • Ridges on the sides of the box for easy grip when wearing gloves

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service