Skip to Content
Merck
All Photos(1)

Key Documents

CLS3993

Corning® 96 Well Half-Area Microplate

flat bottom, black polystyrene, NBS (non-binding surface), bag of 5, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$172.00

$172.00


Estimated to ship onApril 17, 2025



Select a Size

Change View
25 EA
$172.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black bottom
black polystyrene
flat bottom
round wells
size:
96 wells
well D:
10.7 mm
well volume:
190 μL
well working volume:
25-125 μL

$172.00


Estimated to ship onApril 17, 2025


material

black bottom
black polystyrene
flat bottom
round wells

description

non-binding surface

sterility

non-sterile

feature

lid: no
skirt

packaging

bag of 5
pkg of 5 inner package

manufacturer/tradename

Corning 3993

size

96 wells

well D

10.7 mm

well volume

190 μL

well working volume

25-125 μL

color

black

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.

Features and Benefits

  • Corning® 96 Well Half-Area Microplate is ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Designed to reduce well-to-well crosstalk during luminescent assays.
  • Nonbinding Surface (NBS) treated microplates have a non-ionic hydrophilic well surface, which makes them ideal for minimizing protein binding in homogeneous assays.
  • Conforms to standard microplate footprint and dimensions.
  • Flat bottom is best suited for detection.
  • Lids are supplied separately.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service