Skip to Content
Merck
All Photos(1)

Key Documents

CLS3631

Corning®  96 Well Black Polystyrene Microplate

flat bottom clear, black polystyrene, bag of 25, non-sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$620.00

$620.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 EA
$620.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene
flat bottom clear
polystyrene
round clear wells
size:
96 wells

$620.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

black polystyrene
flat bottom clear
polystyrene
round clear wells

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well standard

packaging

case of 100
bag of 25

manufacturer/tradename

Corning 3631

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

clear

suitability

suitable for (fluorescent/luminescent assays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescent and enabling readings down to 340 nm
  • Designed to reduce well-to-well crosstalk during fluorescent assays
  • Can be used for both top and bottom reading instruments
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200 ng IgG/cm2
  • Without lids, lids supplied separately
  • Recommended working volume of 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service