Skip to Content
Merck
All Photos(2)

Key Documents

CLS3915

Corning® 96 Well Solid Polystyrene Microplate

flat bottom, black polystyrene, plate of 100 (in bags of 25), non-sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$378.00

$378.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 EA
$378.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black bottom
black polystyrene
flat bottom
size:
96 wells

$378.00


Estimated to ship onMay 28, 2025


material

black bottom
black polystyrene
flat bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

plate of 100 (in bags of 25)

manufacturer/tradename

Corning 3915

size

96 wells

well volume

360 μL

working volume

75-200 μL

color

black

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Solid Polystyrene Microplates made of opaque black polystyrene walls display minimal well to well crosstalk and background.
  • Recommended working volume of 75 to 200 μL.
  • Low background fluorescence/or luminescence and minimize light scattering.
  • Not treated; medium binding; hydrophobic surface; enable biomolecules binding through passive interactions.
  • Applicable for the immobilization antibodies with hydrophobic regions.
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm².
  • Conforms to standard microplate footprint and dimensions.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service