Skip to Content
Merck
All Photos(1)

Key Documents

211-500

Sigma-Aldrich

Endothelial Cell Growth Medium (500 ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Pricing and availability is not currently available.

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media
Endothelial Cell Growth Medium (500 ml) is an all-in-one ready-to-use. Optimized and best-suited for angiogenesis and other physiological studies.

Application

Endothelial Cell Growth Medium (500 ml) has been used to culture human aortic endothelial cells[1][2] and human umbilical vein endothelial cells.[3][4]

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaya Sriram et al.
International journal of environmental research and public health, 15(10) (2018-10-24)
Airborne particulate matter (PM) exposure remains the leading environmental risk factor for disease globally. Interventions to mitigate the adverse effects of PM are required, since there is no discernible threshold for its effects, and exposure reduction approaches are limited. The
Robin Simsa et al.
PloS one, 14(8), e0220743-e0220743 (2019-08-06)
Decellularization of blood vessels is a promising approach to generate native biomaterials for replacement of diseased vessels. The decellularization process affects the mechanical properties of the vascular graft and thus can have a negative impact for in vivo functionality. The
Yang Li et al.
Life sciences, 232, 116590-116590 (2019-06-23)
Endothelial cell (EC) apoptosis is fundamental for the pathophysiology of atherosclerosis, in which microRNAs (miRNAs) emerge as critical regulators. miR-122 has been shown to regulate the apoptosis of various cell types, however, whether miR-122 is associated with atherosclerosis and EC
Angel Tan et al.
Biomaterials science, 7(8), 3460-3470 (2019-07-04)
In the research field of nanostructured systems for biomedical applications, increasing attention has been paid to using biomimetic, dynamic cellular models to adequately predict their bio-nano behaviours. This work specifically evaluates the biointeractions of nanostructured lipid-based particles (cubosomes) with human
Ekramy Elmorsy et al.
Toxicology in vitro : an international journal published in association with BIBRA, 44, 154-163 (2017-07-18)
Antidepressants are commonly employed for the treatment of major depressive disorders and other psychiatric conditions. We investigated the relatively acute cytotoxic effects of three commonly prescribed antidepressants: fluoxetine, sertraline, and clomipramine on rat primary blood brain barrier endothelial cells over

Articles

Medium 199 supports non-transformed cell cultivation in virology and vaccine production, offering broad species applicability.

Protocols

Technical information for working with human aortic endothelial cells including thawing, subculturing and cryopreservation. Be sure to wear face protection mask and gloves when retrieving cryovials from the liquid nitrogen storage tank. The dramatic temperature change from the tank to the room could cause any trapped liquid nitrogen in the cryovials to burst and cause injury.

Questions

1–4 of 4 Questions  
  1. Does the medium contain VEGF? If so, can you specify the concentration?

    1 answer
    1. This medium does not contain VEGF.

      Helpful?

  2. If I use this medium, do I need to use tissue coated treated flasks with gelatin? Does this medium need to be supplemented with heparin or is it already included?

    1 answer
    1. This medium is a complete, ready-to-use product; no supplementation is required. This formulation contains hydrocortisone, heparin and other growth factors needed to support Endothelial Cell growth. Unless a suspension culture is required, the culture vessel should be tissue culture treated. The additional use of attachment matrices such as gelatin, would be dependent on the requirements of the specific cell line or type.

      Helpful?

  3. Can you please provide the full composition of the medium with the concentrations?

    1 answer
    1. This item is a product of Cell Applications. The formulation is considered proprietary. However, this medium does include glucose at 1g/L, 2% Fetal Bovine Serum, as well as Penicillin, streptomycin, and Amphotericin B. Some salt components have also been published in the Safety Data Sheet.

      Helpful?

  4. Do you sell glucose-free ECGM? I needed it for HUVEC cells. 0mM glucose.

    1 answer
    1. This product is not a glucose-free medium. The formulation contains 5.78 mM glucose which is 5.55 mM after all the supplements are added. Unfortunately, a glucose-free ECGM medium is not currently offered.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service