Skip to Content
Merck
All Photos(1)

Key Documents

CLS3571BC

Corning® 384 well microplate, low volume

Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, with generic barcode, lid, pack of 10

Synonym(s):

384 multiwell plates

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$871.00

$871.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$871.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
black bottom
black polystyrene
flat bottom
size:
384 wells

$871.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

black bottom
black polystyrene
flat bottom

sterility

sterile

feature

lid
skirt
with generic barcode

packaging

pack of 10

manufacturer/tradename

Corning 3570BC

size

384 wells

wells

384 wells

working volume

20-80 μL

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service