Skip to Content
Merck
All Photos(1)

Key Documents

CLS3570

Corning® 384 well microplate, low flange

Tissue Culture (TC)-treated surface, white polystyrene, flat bottom, sterile, lid, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$303.00
50 EA
$359.00

$303.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 EA
$303.00
50 EA
$359.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
flat bottom
polystyrene
square white wells
white bottom
white polystyrene
size:
384 wells

$303.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

flat bottom
polystyrene
square white wells
white bottom
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3570

maximum volume

112 μL
112 μL

size

384 wells

working volume

80 μL
80 μL

color

white

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 384 well microplate, low flange with lid polystyrene white flat bottom plates minimize well-to-well crosstalk during luminescent assays.
  • Matrix active group and tissue culture-treated for optimal cell attachment.
  • Conforms to standard microplate footprint and dimensions.
  • Ideal for today′s more sensitive assays and plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service