Skip to Content
Merck
All Photos(1)

Key Documents

CLS3365

Corning® 96 Well Storage Microplate

round bottom clear, polypropylene, bag of 25 ×, non-sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$293.00

$293.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
100 EA
$293.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
polypropylene
round bottom clear
size:
96 wells

$293.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polypropylene
round bottom clear

sterility

non-sterile

feature

lid: no

manufacturer/tradename

Corning 3365

packaging

bag of 25 ×

size

96 wells

well volume

330 μL

well working volume

75-200 μL

color

clear

suitability

suitable for (general storage, liquid handling; automated applications)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Corning® 96 well microplates support various assay and storage application requirements.
  • Features uniform skirt heights for the greater robotic gripping surface.
  • Resistant to most solvents including DMSO, ethanol, methanol.
  • RNase-/DNase-free.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service