CLS11650
Corning® closed system bottle
capacity 150 mL, polystyrene, male Luer Lock, sterile
Synonym(s):
Closed System Bottle, Corning System Bottle
Select a Size
$1,780.00
Select a Size
About This Item
$1,780.00
Recommended Products
material
clear bottle
high-density polyethylene cap
polystyrene
round bottle
wide-mouth bottle
sterility
sterile
feature
cap
male Luer Lock
packaging
pkg of 8 ea
manufacturer/tradename
Corning 11650
bottle neck diam.
~42 mm
capacity
150 mL
Related Categories
General description
The preassembled transfer caps have two ports.
One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
- Made from optically clear polystyrene
- Sterilized to SAL 10-6
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)