CLS11460
Corning® Erlenmeyer flasks
capacity 2 L, sterile, flat bottom, 2 port, no vent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
flat bottom
sterility
sterile
feature
cap: no
packaging
pack of 4 ea
manufacturer/tradename
Corning 11460
I.D.
1/8 in.
capacity
2 L
pore size
0.2 μm
fitting
male Luer for lock
Related Categories
Other Notes
Browse all Erlenmeyer flasks
Legal Information
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service