Skip to Content
Merck
All Photos(1)

Key Documents

CAS9GEMP

Sigma-Aldrich

Cas9 Geminin plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 μG
$299.00

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

Select a Size

Change View
1 μG
$299.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

$299.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 4 to 6 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

Promoter

Promoter name: EF1-alpha

reporter gene

GFP

selection

ampicillin

shipped in

dry ice

storage temp.

−20°C

General description

This product is an expression plasmid that utilizes the EF1a promoter for strong transient expression of a Cas9-GFP-Geminin fusion (EF1a-Cas9-GFP-Geminin) allowing for easy visualization of successful transfection. The Cas9-Geminin expression plasmid is one part of a two part CRISPR system with individual Cas9-Geminin and gRNA expression vectors.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Performing HDR mediated targeted integration in multiple cell lines
  • Creation of knock-in cell lines with promoters, fusion tags or reporters integrated into endogenous genes

Features and Benefits

  • Highly specific and highly active
  • Sequence verified
  • Ready to use purified plasmid DNA

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB. While HDR is absent in G1, NHEJ is active throughout the cell cycle and is largely favored over HDR. Consequently HDR can be increased by directly synchronizing the expression of Cas9 with cell-cycle progression by fusing Cas9 to human Geminin (a protein expressed in S and G2 phases). The Cas9-geminin fusion protein then regulates gene editing by promoting repair during S and G2 phases when homology directed repair (HDR) occurs. The result is creation of double strand breaks in cells at times that are more able to incorporate donor sequences.

Legal Information

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service