Skip to Content
Merck
All Photos(1)

Key Documents

C7495

Sigma-Aldrich

Clofarabine

≥98% (HPLC)

Synonym(s):

(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol, 2-chloro-2′-arabino-fluoro-2′-deoxyadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClFN5O3
CAS Number:
Molecular Weight:
303.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >10 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

Nc1nc(Cl)nc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3F

InChI

1S/C10H11ClFN5O3/c11-10-15-7(13)5-8(16-10)17(2-14-5)9-4(12)6(19)3(1-18)20-9/h2-4,6,9,18-19H,1H2,(H2,13,15,16)/t3-,4+,6-,9-/m1/s1

InChI key

WDDPHFBMKLOVOX-AYQXTPAHSA-N

Application

Clofarabine has been used in a cell viability assay to analyze the sensitivity of the isogenic cell lines towards clofarabine. It is also used to study the interaction of anticancer drug clofarabine with human serum albumin and human α-1 acid glycoprotein.

Biochem/physiol Actions

Clofarabine is a purine nucleoside antimetabolite. Clofarabine is toxic to nondividing lymphocytes and monocytes.
Clofarabine is a second-generation nucleoside analog, used in cancer treatments. Clofarabine is metabolized to 5′-triphosphate and is known to block DNA synthesis. The human equilibrative nucleoside transporters (hENT1 and hENT2) and human concentrative nucleoside transporter (hCNT2 and hCNT3) mediates clofarabine transport into the cell. It also serves as a substrate for mitochondrial deoxyguanosine kinase. Clofarabine is an inhibitor of ribonucleotide reductase. It resists the phosphorolytic cleavage catalyzed by purine nucleoside phosphorylase of bacterias. Clofarabine also withstands deamination by adenosine deaminase.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Results of a phase 1-2 study of clofarabine in combination with cytarabine (ara-C) in relapsed and refractory acute leukemias
Faderl S, et al.
Blood, 105(3), 940-947 (2005)
Michał Gorzkiewicz et al.
Macromolecular rapid communications, 40(15), e1900181-e1900181 (2019-05-29)
Poly(propyleneimine) glycodendrimers are proposed as nanocarriers for triphosphate forms of anticancer adenosine analogues to improve the efficiency of chemotherapy and to overcome drug resistance mechanisms. This approach has proven successful for fludarabine administration-an autonomous way of cellular entry of a
Nobuko Hijiya et al.
Pediatric blood & cancer, 59(3), 417-422 (2012-02-23)
Clofarabine is a second-generation purine nucleoside analog and has significant anti-leukemic activity as a single agent. It is approved by the United States Food and Drug Administration (FDA) for the treatment of relapsed or refractory acute lymphoblastic leukemia (ALL) in
Stratification of nucleoside analog chemotherapy using 1-(2?-deoxy-2?-18F-fluoro-beta-D-arabinofuranosyl) cytosine and 1-(2?-deoxy-2?-18F-fluoro-beta-L-arabinofuranosyl)-5-methylcytosine PET
Lee JT, et al.
Journal of Nuclear Medicine, 53(2), 275-275 (2012)
Modified Nucleosides: in Biochemistry, Biotechnology and Medicine, 639-639 (2008)

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service