Key Documents
BAF148480000
Fillpour® Dialysis Tube Funnels
5, 9 and 13.5 mm Diameter Steps, 10 pak of
Select a Size
10 EA
$126.00
$126.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$126.00
About This Item
UNSPSC Code:
41123100
NACRES:
NB.24
$126.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
manufacturer/tradename
Bel-Art F14848-0000
Scienceware®
packaging
10 pak of
Related Categories
General description
To use, cut dialysis tubing to length and knot one end. Slip the open end onto the tube funnel and secure with included rubber band or Scienceware® silicone rubber loops. A captive snap on cover has a small writing area for easy identification. Steam autoclavable at 121 deg C (250 deg F). 10 per bag.
Supports dialysis tubes on edge of vessel and allows easy filling.
Features and Benefits
  • Durable polypropylene construction
  • Allows convenient handling of dialysis bags
  • Three tube steps fit various sizes of dialysis tubing
  • Hanger has three different notches to fit the edge of any vessel
Legal Information
Fillpour is a registered trademark of SP Scienceware, Division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)