Skip to Content
Merck
All Photos(1)

Key Documents

D0655

Sigma-Aldrich

Dialysis tubing, high retention seamless cellulose tubing

avg. flat width 40 mm (1.6 in.), MWCO 12400, 99.99% retention

Sign Into View Organizational & Contract Pricing

Select a Size

100 FT
$561.00

$561.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details



Select a Size

Change View
100 FT
$561.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

$561.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


avg. diam.

25 mm , when full

avg. flat width

40 mm (1.6 in.)

capacity

~150 mL/ft

pore size

12400 MWCO, 99.99% retention

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Retains >99% of cytochrome c (M.W. 12,400) in solution over a 10-hour period.

Packaging

Supplied in rolls, dry

Quality

May contain glycerin and sulfur compounds in more than trace amounts; washing instructions are included.

Preparation Note

Removal of glycerol included as a humectant can be accomplished by washing the tubing in running water for 3-4 hours. Removal of sulfur compounds can be accomplished by treating the tubing with a 0.3% (w/v) solution of sodium sulfide at 80°C for 1 minute. Wash with hot water (60°C) for 2 minutes, followed by acidification with a 0.2% (v/v) solution of sulfuric acid, then rinse with hot water to remove the acid. This tubing will retain most proteins of molecular weight 12,000 or greater.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Luísa Tomás et al.
Frontiers in microbiology, 10, 2917-2917 (2020-01-11)
Pneumocystis jirovecii pneumonia (PcP) is a major human immunodeficiency virus (HIV)-related illness, rising among immunocompromised non-HIV patients and in developing countries. Presently, the diagnosis requires respiratory specimens obtained through invasive and costly techniques that are difficult to perform in all

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service