Skip to Content
Merck
All Photos(1)

Key Documents

B0434

Sigma-Aldrich

BIM-23027 trifluoroacetate salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H54N8O7
Molecular Weight:
794.94
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

solubility

H2O: soluble

storage temp.

−20°C

Gene Information

human ... SSTR2(6752)
mouse ... SSTR2(20606)
rat ... SSTR2(54305)

Amino Acid Sequence

MeAla-Tyr-Trp-Lys-Abu-Phe

Biochem/physiol Actions

sst2 somatostatin receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service