Skip to Content
Merck
All Photos(1)

Key Documents

B-090

Supelco

N-Butylpentylone hydrochloride solution

1 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO3 · HCl
CAS Number:
Molecular Weight:
313.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C(NCCCC)CCC)C1=CC(OCO2)=C2C=C1.Cl

InChI key

ZQFVGUBLUFAGTB-UHFFFAOYSA-N

General description

This Certified Spiking Solution® is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology to urine drug testing. N-Butylpentylone is a stimulant drug of the cathinone class which has been reported as a novel designer drug.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service