Skip to Content
Merck
All Photos(1)

Key Documents

46920

Sigma-Aldrich

Fmoc N-hydroxysuccinimide ester

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

9-Fluorenylmethyl N-succinimidyl carbonate, N-(9-Fluorenylmethoxycarbonyloxy)succinimide, Fmoc-OSu

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$115.00
25 G
$271.00
100 G
$518.00

$115.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$115.00
25 G
$271.00
100 G
$518.00

About This Item

Empirical Formula (Hill Notation):
C19H15NO5
CAS Number:
Molecular Weight:
337.33
Beilstein:
3569540
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$115.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Fmoc N-hydroxysuccinimide ester, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

mp

150-153 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc
imide

storage temp.

2-8°C

SMILES string

O=C(OCC1c2ccccc2-c3ccccc13)ON4C(=O)CCC4=O

InChI

1S/C19H15NO5/c21-17-9-10-18(22)20(17)25-19(23)24-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16H,9-11H2

InChI key

WMSUFWLPZLCIHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids in high yield. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It has been employed in the synthesis of glycopeptides.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved purities for Fmoc-amino acids from Fmoc-ONSu.
R C Milton et al.
International journal of peptide and protein research, 30(3), 431-432 (1987-09-01)
A. Paquet
Canadian Journal of Chemistry, 60, 976-976 (1982)
Occupational airborne allergic contact dermatitis from succinimidyl carbonates.
J F Fowler et al.
Contact dermatitis, 45(1), 38-38 (2001-06-26)
Huaimin Wang et al.
Scientific reports, 5, 16680-16680 (2015-11-18)
Biocompatible peptide-based supramolecular hydrogel has recently emerged as a new and promising system for biomedical applications. In this work, Rhodamine B is employed as a new capping group of self-assembling peptide, which not only provides the driving force for supramolecular
Ning Wang et al.
Carbohydrate research, 411, 37-41 (2015-05-15)
Monoglucosylated high-mannose-type glycan (Glc1Man9GlcNAc2: G1M9) is well-known as a key glycoform in the glycoprotein folding process, which is specifically recognized by lectin chaperones calnexin (CNX) and calreticulin (CRT) in the endoplasmic reticulum (ER). In this work, we developed an efficient

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service