Skip to Content
Merck
All Photos(2)

Key Documents

AV40225

Sigma-Aldrich

Anti-EXOSC10 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Exosome component 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

97 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXOSC10(5394)

General description

Exosome complex (RNAse complex), a complex of 3′ -→ 5′ exoribonucleases, is a multi-protein complex that degrades various types of ribonucleic acids (RNA). Exosome component 10 (EXOSC10, PMSCL2, "polymyositis/scleroderma autoantigen 2, 100kDa) is an exoribonuclease that shares sequence similarity to budding yeast Rrp6 and is proposed to catalyze 3′-to-5′ exoribonuclease activity on a variety of nuclear transcripts including ribosomal RNA subunits, RNA that has been poly-adenylated by TRAMP, as well as other nuclear RNA transcripts destined for processing and/or destruction.

Specificity

Anti-EXOSC10 polyclonal antibody reacts with human, mouse, rat, and bovine polymyositis/scleroderma autoantigen 2, 100kDa (Rrp6) proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human EXOSC10

Application

Anti-EXOSC10 polyclonal antibody is used to tag polymyositis/scleroderma autoantigen 2, 100kDa for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the possible roles of polymyositis/scleroderma autoantigen 2, 100kDa (Rrp6) in the function of specific exosomes.

Biochem/physiol Actions

EXOSC10 contains 1 HRDC domain and 1 3′-5′ exonuclease domain. Antibodies against PM/SCL are found in patients with polymyositis and/or scleroderma.

Sequence

Synthetic peptide located within the following region: FAGNSKSKVSSQFDPNKQTPSGKKCIAAKKIKQSVGNKSMSFPTGKSDRG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service