Skip to Content
Merck
All Photos(1)

Key Documents

890810P

Avanti

18:0 DDAB

Avanti Research - A Croda Brand

Synonym(s):

Dimethyldioctadecylammonium (Bromide Salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$229.00
200 MG
$397.00
1 G
$985.00

$229.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$229.00
200 MG
$397.00
1 G
$985.00

About This Item

Empirical Formula (Hill Notation):
C38H80NBr
CAS Number:
Molecular Weight:
630.95
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$229.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 g (890810P-1g)
pkg of 1 × 200 mg (890810P-200mg)
pkg of 1 × 25 mg (890810P-25mg)
pkg of 1 × 500 mg (890810P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCCCC)(C)C.[Br-]

InChI

1S/C38H80N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1

InChI key

PSLWZOIUBRXAQW-UHFFFAOYSA-M

General description

18:0 DDAB is cationic lipid[1] and a quaternary ammonium salt.[2] The hydrocarbon tails are saturated unlike the ionizable lipids.[1]

Application

18:0 DDAB is suitable for the preparation of lipid-nanoparticles.[1] It has also been used as an adjuvant and combined with monophosphoryl lipid A (MPL) for immunizing mice.[3]

Biochem/physiol Actions

18:0 DDAB is a surface-active molecule, used as an adjuvant to induce systemic immune response. It is non-toxic.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (890810P-1g)
20 mL Clear Glass Screw Cap Vial (890810P-500mg)
5 mL Amber Glass Screw Cap Vial (890810P-200mg)
5 mL Amber Glass Screw Cap Vial (890810P-25mg)

Other Notes

For R&D use only. Not for drug, household, or other uses.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipophilic quaternary ammonium salt acts as a mucosal adjuvant when co-administered by the nasal route with vaccine antigens
Klinguer C, et al.
Vaccine, 19(30), 4236-4244 (2001)
Ana Cristina Norberto Oliveira et al.
ACS applied materials & interfaces, 6(9), 6977-6989 (2014-04-10)
This study describes a novel liposomal formulation for siRNA delivery, based on the mixture of the neutral lipid monoolein (MO) and cationic lipids of the dioctadecyldimethylammonium (DODA) family. The cationic lipids dioctadecyldimethylammonium bromide (DODAB) and chloride (DODAC) were compared in
Hong Yu et al.
Infection and immunity, 78(5), 2272-2282 (2010-03-17)
Major impediments to developing a Chlamydia vaccine lie in identifying immunologically relevant T-cell antigens and delivery in a manner to stimulate protective immunity. Using an immunoproteomic approach, we previously identified three immunodominant Chlamydia T-cell antigens (PmpG-1, PmpE/F-2, and RplF). Because
Lipid nanoparticles for delivery of messenger RNA to the back of the eye
Patel S, et al.
J. Controlled Release, 303, 91-100 (2019)
Hong Yu et al.
Infection and immunity, 80(4), 1510-1518 (2012-02-01)
Major impediments to a Chlamydia vaccine lie in discovering T cell antigens and polarizing adjuvants that stimulate protective immunity. We previously reported the discovery of three T cell antigens (PmpG, PmpF, and RplF) via immunoproteomics that elicited protective immunity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service