Skip to Content
Merck
All Photos(4)

Key Documents

A6926

Sigma-Aldrich

3-Amino-9-ethylcarbazole

chromogenic, tablet

Synonym(s):

9-Ethylcarbazol-3-amine, AEC

Sign Into View Organizational & Contract Pricing

Select a Size

5 TABS
$75.50
50 TABS
$285.00
100 TABS
$519.00

$75.50


Estimated to ship onMay 24, 2025



Select a Size

Change View
5 TABS
$75.50
50 TABS
$285.00
100 TABS
$519.00

About This Item

Empirical Formula (Hill Notation):
C14H14N2
CAS Number:
Molecular Weight:
210.27
Beilstein:
174969
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$75.50


Estimated to ship onMay 24, 2025


Product Name

3-Amino-9-ethylcarbazole, tablet

form

tablet

Quality Level

solubility

DMF: 1 tablet/5 mL

SMILES string

CCn1c2ccccc2c3cc(N)ccc13

InChI

1S/C14H14N2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2,15H2,1H3

InChI key

OXEUETBFKVCRNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Amino-9-ethylcarbazole is a chromogen

Specificity

3-Amino-9-ethylcarbazole is a peroxidase substrate suitable for use in immunoblotting and immunohistochemical staining procedures. This substrate produces an insoluble end product that is red in color and can be observed visually.

Application

3-Amino-9-ethylcarbazole (AEC) has been used as a chromogen in immunohistochemistry to visualize the cells.[1] It has also been used as a substrate in enzyme-linked immune absorbent spot (ELISpot).[2]

Quantity

Contains 20 mg substrate per tablet.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Human parainfluenza virus-3 can be targeted by rapidly ex vivo expanded T lymphocytes
McLaughlin LP, et al.
Cytotherapy, 18(12), 1515-1524 (2016)
Cristan Herbert et al.
Respirology (Carlton, Vic.), 20(8), 1206-1212 (2015-08-21)
Exacerbations of allergic asthma are often triggered by respiratory viral infections. We have previously shown that in a T-helper type 2 (Th2)-biased cytokine environment, mouse and human airway epithelial cells (AEC) exhibit increased expression of pro-inflammatory and anti-viral genes in
G P Harvey et al.
Journal of periodontal research, 48(2), 252-261 (2012-09-18)
The presence of citrullinated proteins, and peptidylarginine deiminase types -2 (PAD-2) and -4 (PAD-4) in periodontal tissues, determine the presence of anti-cyclic citrullinated protein antibodies (anti-CCP) in gingival crevicular fluid (GCF) and compare the expression of these proteins between inflamed
Nicolas Kluger et al.
Clinical immunology (Orlando, Fla.), 158(2), 212-220 (2015-03-26)
Gastrointestinal dysfunction is a disabling manifestation of APECED possibly related to an autoimmune intestinal aggression. We evaluated its features in a cohort of 31 Finnish patients. The most frequent manifestations were constipation (48%), diarrhea, dysphagia and retrosternal pain (45%). AADC
Iresha N Ambepitiya Wickramasinghe et al.
PloS one, 10(6), e0128893-e0128893 (2015-06-04)
The initial interaction between viral attachment proteins and the host cell is a critical determinant for the susceptibility of a host for a particular virus. To increase our understanding of avian pathogens and the susceptibility of poultry species, we developed

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service