Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Amino-9-ethylcarbazole
Synonym(s):
9-Ethylcarbazol-3-amine, AEC
Empirical Formula (Hill Notation):
C
14
H
14
N
2
CAS Number:
132-32-1
Molecular Weight:
210.27
EC Number:
205-057-7
Products
Product Category
substrates (2)
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A5754
3-Amino-9-ethylcarbazole,
≥95% (HPLC), powder
Expand
View Pricing
A6926
3-Amino-9-ethylcarbazole,
chromogenic, tablet
Expand
View Pricing