Skip to Content
Merck
All Photos(1)

Documents

815-500

Sigma-Aldrich

Human Mammary Epithelial Cell Growth Medium (500ml)

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

All-in-one ready-to-use media

Application

Human Mammary Epithelial Cell Growth Medium (500ml) has been used to culture MCF10A cells and human primary mammary epithelial cells.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whitney S Henry et al.
Cancer research, 77(3), 790-801 (2016-12-13)
Despite the high incidence of oncogenic mutations in PIK3CA, the gene encoding the catalytic subunit of PI3K, PI3K inhibitors have yielded little clinical benefit for breast cancer patients. Recent epidemiologic studies have suggested a therapeutic benefit from aspirin intake in
Andrew Alsterda et al.
Frontiers in oncology, 11, 654940-654940 (2021-06-08)
The endoplasmic reticulum (ER) regulates protein folding, post-translational modifications, lipid synthesis, and calcium signaling to attenuate the accumulation of misfolded proteins causing ER stress and maintains cellular homeostasis. The tumor microenvironment is rich in soluble cytokines, chemokines, growth, and angiogenic
Kara McNair et al.
Cancer biology & therapy, 20(3), 349-367 (2018-11-08)
Expression of the tumour suppressor Deleted in Colorectal Cancer (DCC) and the related protein neogenin is reduced by the mammalian serine protease chymotrypsin or the bacterial serine protease subtilisin, with increased cell migration. The present work examines whether these actions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service