Skip to Content
Merck
All Photos(1)

Key Documents

46939

Sigma-Aldrich

Fluorescein diacetate 5-maleimide

suitable for fluorescence

Synonym(s):

5-Maleimido-fluorescein diacetate, Fluorescein-5-maleimide diacetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$560.00

$560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$560.00

About This Item

Empirical Formula (Hill Notation):
C28H17NO9
CAS Number:
Molecular Weight:
511.44
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32

$560.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

solubility

DMF: soluble
DMSO: soluble
dioxane: soluble

fluorescence

λex 492 nm; λem 520 nm in 0.1 M phosphate pH 7.0 (after derivatization with 2-mercaptoethanol and after cleavage by esterase)

suitability

corresponds for coupling to thiols
suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5cc(ccc45)N6C(=O)C=CC6=O)c1

InChI

1S/C28H17NO9/c1-14(30)35-17-4-7-21-23(12-17)37-24-13-18(36-15(2)31)5-8-22(24)28(21)20-6-3-16(11-19(20)27(34)38-28)29-25(32)9-10-26(29)33/h3-13H,1-2H3

InChI key

WZBJWHQRQDEKOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescein diacetate 5-maleimide can be used as a substrate to prepare fluorescent Fe3O4 superparamagnetic nanoparticles (SPMNP) applicable in the subcellular fractionation process.[1] It is also used in the surface modification of glass substrates to alter the surface properties.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Formation of conjugates with immunoreactant for photoluminometric immunoassay by fluorescent energy transfer[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Lakowicz et al.
Eur. Pat. Appl. 552108, CA120, 101282 (1993)
Preparation and characterization of alkyl-thiols monolayer on glass substrates by microcontact printing
Abdullah NH, et al.
AAPS PharmSci, 10, 9538-9543 (2015)
Surface functionalization dependent subcellular localization of Superparamagnetic nanoparticle in plasma membrane and endosome
Raj Deepak BTG and Khan NA
Nano convergence, 5, 4-4 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service