Skip to Content
Merck
All Photos(1)

Key Documents

28562

Sigma-Aldrich

Atto 488 maleimide

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Atto 488

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$774.00

$774.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$774.00

About This Item

MDL number:
UNSPSC Code:
12352111
NACRES:
NA.32

$774.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (degree of coupling )

manufacturer/tradename

ATTO-TEC GmbH

λ

in methanol: water (1:1) (with 0.1% perchloric acid)

UV absorption

λ: 501-506 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 488 Maleimide is a hydrophilic fluorescent label with high molecular absorption (90.000) and quantum yield (0.80). The fluorescence activity is excited at a maximum of 501nm, and the maximum emission is 523nm. Atto 488 Maleimide is optimized for argon laser excitation and characterized by high photostability. Hydrolysis of maleimides to a mixture of isomeric nonreactive maleamic acids can compete significantly with thiol modification, particularly above pH 8. Post coupling to a substrate, the label carries a net electrical charge of -1.

Application

Atto 488 Maleimide is used for labelling amines, which usually requires a higher pH than the reaction of maleimides with thiols. It is a well-suited fluorescent label for thiol groups.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of fluorescent nanostructures in biological systems by means of spectral position determination microscopy (SPDM).
Muller, P., et al. et al.
Current Microscopy Contributions to Advances in Science and Technology, 1, 3-12 (2012)
Mohd A Mohd Ridzuan et al.
PloS one, 7(3), e33845-e33845 (2012-04-06)
An actomyosin motor complex assembled below the parasite's plasma membrane drives erythrocyte invasion by Plasmodium falciparum merozoites. The complex is comprised of several proteins including myosin (MyoA), myosin tail domain interacting protein (MTIP) and glideosome associated proteins (GAP) 45 and
Lucia De Rosa et al.
Organic & biomolecular chemistry, 10(2), 273-280 (2011-11-11)
In the last few years, the use of labeled proteins has significantly expanded in the life sciences. Now, labeled proteins are indispensable tools for a wide spectrum of biophysical and chemical biology applications. In particular, the quest for more sophisticated
Jonas K Hannestad et al.
ACS nano, 7(1), 308-315 (2012-12-12)
We use single-molecule fluorescence microscopy to monitor individual hybridization reactions between membrane-anchored DNA strands, occurring in nanofluidic lipid monolayer films deposited on Teflon AF substrates. The DNA molecules are labeled with different fluorescent dyes, which make it possible to simultaneously
Tilak Jain et al.
Journal of structural biology, 179(1), 68-75 (2012-05-10)
Over the last three decades, Cryo-TEM has developed into a powerful technique for high-resolution imaging of biological macromolecules in their native vitrified state. However, the method for vitrifying specimens onto EM grids is essentially unchanged - application of ∼3 μL

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service